Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026860.1 | 2 | 869 | |||
GenBank mRNA | AK289375.1 | - | 2 | 753 | ||
BC008632.1 | - | 2 | 869 | |||
BC039614.1 | - | 2 | 869 |