Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VAC14-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_034083.2 | 6 | 2136 | |||
GenBank mRNA | AK095020.1 | - | 6 | 2062 | ||
BC035152.1 | - | 5 | 4764 |