Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC440602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108042.2 | 1 | 80 | |||
GenBank mRNA | BC042100.1 | - | 1 | 80 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ049505.1 | - | 78 | Chr. - - [-] on Build |