Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01220 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038421.1 | 3 | 423 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CN263935.1 | - | 3 | 421 | Chr. 14 - 75294407 - 75296638 [+] on Build GRCh38 |