Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928937 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110095.1 | 1 | 171 | |||
GenBank mRNA | BC043545.1 | - | 1 | 171 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI829119.1 | - | 1 | 170 | Chr. 12 - 99093359 - 99104910 [+] on Build GRCh38 |