Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101060391 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_136151.1 | 3 | 713 | |||
GenBank mRNA | CD519625.1 | - | 2 | 511 | ||
DB544287.1 | - | 4 | 455 | |||
DB545731.1 | - | 4 | 467 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CD557060.1 | - | 2 | 493 | Chr. 2 - 949632 - 950266 [-] on Build GRCh38 |