Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100507657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_134581.1 | 1 | 3460 | |||
GenBank mRNA | CR627208.1 | - | 1 | 3459 | ||
CR936633.1 | - | 1 | 3459 |