Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MYCNOS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026766.1 | 2 | 282 | |||
NR_110230.1 | 2 | 282 | ||||
GenBank mRNA | BC002892.2 | - | 2 | 141 | ||
S49953.1 | - | 2 | 282 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX279550.1 | - | 1 | 231 | Chr. 2 - 15940121 - 15941238 [-] on Build GRCh38 |