Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928008 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120539.1 | 3 | 275 | |||
GenBank mRNA | U80769.1 | - | 3 | 287 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | H85304.1 | - | 3 | 287 | Chr. 11 - 9839142 - 9929035 [+] on Build GRCh38 |