Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927438 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110224.1 | 5 | 1714 | |||
GenBank mRNA | AK097952.1 | - | 5 | 1709 |