Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf99 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195032.1 | NP_001181961.1 | 3 | 599 | ||
NM_001302839.1 | NP_001289768.1 | 3 | 933 | |||
NM_001302840.1 | NP_001289769.1 | 4 | 694 | |||
NM_001302841.1 | NP_001289770.1 | 3 | 593 | |||
XM_005266788.4 | XP_005266845.1 | 3 | 940 | |||
XM_017010142.1 | XP_016865631.1 | 3 | 530 | |||
XM_017010143.1 | XP_016865632.1 | 3 | 607 | |||
XM_017010144.1 | XP_016865633.1 | 3 | 602 | |||
XM_017010145.1 | XP_016865634.1 | 2 | 505 | |||
XM_017010146.1 | XP_016865635.1 | 2 | 500 | |||
XM_017010147.1 | XP_016865636.1 | 3 | 590 |