Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100130111 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135221.1 | 2 | 893 | |||
GenBank mRNA | AK096061.1 | - | 2 | 838 |