Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM161B-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_039993.1 | 4 | 671 | |||
NR_039994.2 | 4 | 629 | ||||
NR_039995.1 | 3 | 549 | ||||
NR_105019.1 | 8 | 1050 | ||||
NR_105020.1 | 3 | 613 | ||||
GenBank mRNA | BC030122.2 | - | 4 | 615 | ||
BC047059.1 | - | 8 | 1042 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG700933.1 | - | 4 | 620 | Chr. 5 - 88268938 - 88436639 [+] on Build GRCh38 |