Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC102723385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_119379.1 | 3 | 298 | |||
NR_119380.1 | 2 | 230 | ||||
NR_119381.1 | 2 | 200 | ||||
NR_119382.1 | 2 | 255 | ||||
GenBank mRNA | FY212114.1 | - | 3 | 249 | ||
FY213069.1 | - | 2 | 181 | |||
FY213574.1 | - | 2 | 151 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | FY213705.1 | - | 3 | 250 | Chr. 16 - 19062862 - 19067635 [-] on Build GRCh38 |