Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSC22D1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038381.1 | 3 | 2070 | |||
GenBank mRNA | BC040675.2 | - | 3 | 2073 |