Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MORC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BM683680.1 | - | 1 | 133 |
Gene Symbol: MORC2-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026920.1 | 2 | 726 | |||
GenBank mRNA | AK093120.1 | - | 2 | 625 | ||
AK098685.1 | - | 1 | 107 | |||
BC042980.1 | - | 3 | 865 | |||
BC065553.1 | - | 2 | 619 | |||
BC117160.1 | - | 1 | 361 | |||
BC117186.1 | - | 1 | 361 |