Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101928441 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120471.1 | 5 | 1600 | |||
GenBank mRNA | AK058180.1 | - | 5 | 1588 |