Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3BP5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046084.1 | 1 | 213 | |||
GenBank mRNA | AL133111.1 | - | 1 | 213 |
Gene Symbol: SH3BP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA429664.1 | - | 1 | 55 |