Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GACAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120598.1 | 2 | 756 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX095757.1 | - | 2 | 750 | Chr. 18 - 8695904 - 8707615 [-] on Build GRCh38 |