Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf197 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001302813.1 | NP_001289742.1 | 3 | 2058 | ||
NM_001302815.1 | NP_001289744.1 | 4 | 1747 | |||
NM_173644.2 | NP_775915.1 | 4 | 2138 | |||
XM_017027828.1 | XP_016883317.1 | 3 | 10863 | |||
XM_017027829.1 | XP_016883318.1 | 4 | 8525 | |||
XM_017027830.1 | XP_016883319.1 | 3 | 8390 | |||
XM_017027831.1 | XP_016883320.1 | 4 | 10209 | |||
XM_017027832.1 | XP_016883321.1 | 4 | 2262 | |||
GenBank mRNA | AK091179.1 | - | 4 | 2138 | ||
AY358539.1 | - | 1 | 3847 |