Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC101927286 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120324.1 | 1, 2 | 240 | |||
GenBank mRNA | BC038379.1 | - | 1, 2 | 240 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BI756744.1 | - | 1, 2 | 239 | Chr. 15 - 97371289 - 97521811 [-] on Build GRCh38 |