Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRCAT47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_131947.1 | 4 | 471 | |||
NR_131948.1 | 3 | 357 | ||||
GenBank mRNA | AK093002.1 | - | 3 | 357 | ||
BG166652.1 | - | 3 | 341 | |||
BG171753.1 | - | 308 | ||||
CR739737.1 | - | 2 | 294 | |||
CR748417.1 | - | 4 | 435 | |||
DA944068.1 | - | 3 | 357 | |||
DA950319.1 | - | 3 | 354 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DA879998.1 | - | 4 | 457 | Chr. 16 - 80830494 - 80870520 [-] on Build GRCh38 |