Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100506603 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_104183.1 | 3 | 852 | |||
NR_104184.1 | 2 | 693 | ||||
GenBank mRNA | AK125727.1 | - | 2 | 692 | ||
BI907467.1 | - | 3 | 280 | |||
DA369488.1 | - | 2 | 488 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY307999.1 | - | 1 | 321 | Chr. 14 - 76782541 - 76783017 [-] on Build GRCh38 |