Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEMA3F-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135301.1 | 4 | 2467 | |||
GenBank mRNA | AK125500.1 | - | 4 | 2470 |