Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HIPK1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110725.1 | 1 | 351 | |||
GenBank mRNA | BC036361.1 | - | 1 | 351 | ||
BC048113.1 | - | 1 | 325 |