Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01525 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_126408.1 | 2 | 328 | |||
GenBank mRNA | BX109912.1 | - | 2 | 274 | ||
DB457076.1 | - | 2 | 328 |