Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR17HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027349.1 | 2 | 185 | |||
NR_027350.1 | 2 | 185 | ||||
GenBank mRNA | AB176707.1 | - | 2 | 185 | ||
BC040320.1 | - | 2 | 185 | |||
BC109081.2 | - | 2 | 63 | |||
BC109082.2 | - | 2 | 63 |
Gene Symbol: MIR18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AB176708.1 | - | LIT3364 oncomir-1 |
2 | 185 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG772153.1 | - | 2 | 190 | Chr. 13 - 91347817 - 91354466 [+] on Build GRCh38 |