Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: B3GALT5-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026542.1 | 3 | 912 | |||
GenBank mRNA | BC119738.2 | - | 3 | 505 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000489821.4 | - | 2 | 619 | Chr. 21 - 39597147 - 39599656 [-] on Build GRCh38 |