Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2610305D13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145078.2 | NP_659544.2 | 5 | 608 | ||
GenBank mRNA | AK011986.1 | - | 2 | 310 | ||
BC032186.1 | - | 1 | 96 | |||
BC058858.1 | - | 5 | 613 | |||
BC138179.1 | - | 2 | 251 | 8861198 | ||
BC145854.1 | - | 2 | 251 | 40131157 |