Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vmn1r51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011683.2 | NP_035813.2 | 3 | 1394 | ||
GenBank mRNA | BC046308.1 | - | 3 | 1394 | 6306893 | |
Y12725.1 | - | 2 | 1364 |
Gene Symbol: Vmn1r42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053221.2 | NP_444451.2 | 1 | 577 | ||
GenBank mRNA | BC125372.1 | - | 1 | 577 | 40129887 |