Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Palmd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023245.3 | NP_075734.3 | 7 | 1480 | ||
GenBank mRNA | AF263246.1 | - | 7 | 1184 | ||
AJ312215.1 | - | 7 | 1203 | |||
AK014531.1 | - | 7 | 1278 | |||
AK136939.1 | - | 7 | 1387 | |||
AK171869.1 | - | 7 | 1211 | |||
AL391037.1 | - | 7 | 1184 | |||
BC010193.1 | - | 7 | 1244 | 3156945 |