Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081684.1 | NP_001075153.1 | 2 | 394 | ||
NM_001081685.1 | NP_001075154.1 | 3 | 537 | |||
NM_175428.3 | NP_780637.2 | 3 | 537 | |||
GenBank mRNA | AK031118.1 | - | 3 | 526 | ||
AK034057.1 | - | 3 | 538 | |||
AK129313.1 | - | 2 | 371 | |||
AK145072.1 | - | 2 | 389 | |||
BC027135.1 | - | 1 | 191 | 5360683 |