Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033444.2 | NP_254279.1 | 5 | 628 | ||
GenBank mRNA | AK088809.1 | - | 5 | 439 | ||
AK150382.1 | - | 5 | 906 | |||
AK150611.1 | - | 5 | 617 | |||
AK153509.1 | - | 5 | 906 | |||
AK167765.1 | - | 5 | 634 | |||
AK168262.1 | - | 5 | 448 | |||
AK186702.1 | - | 3 | 202 | |||
BC004658.1 | - | 5 | 628 | 3498118 |