Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brdt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079873.1 | NP_001073342.1 | 6, 7 | 914 | ||
NM_054054.2 | NP_473395.2 | 6, 7 | 914 | |||
GenBank mRNA | AB208640.1 | - | 4, 5 | 607 | ||
AF358660.1 | - | 6, 7 | 949 | |||
AK077137.1 | - | 6, 7 | 934 | |||
BY726053.1 | - | 2, 3 | 240 |