Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Selm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053267.2 | NP_444497.1 | 4 | 495 | ||
GenBank mRNA | AK018791.1 | - | 4 | 331 | ||
AK035312.1 | - | 4 | 285 | |||
AK046034.1 | - | 4 | 287 | |||
AK079576.1 | - | 2 | 1024 | |||
AY043488.1 | - | 4 | 311 | |||
BC019742.1 | - | 4 | 187 | |||
BC147332.1 | - | 4 | 405 | 9056406 | ||
BC147333.1 | - | 4 | 407 | 9056396 | ||
CK787580.1 | - | 4 | 487 |