Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prosc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039077.2 | NP_001034166.1 | 5 | 401 | ||
NM_001039078.2 | NP_001034167.1 | 4 | 365 | |||
NM_054057.4 | NP_473398.1 | 5 | 401 | |||
GenBank mRNA | AK032308.1 | - | 5 | 351 | ||
AK049905.1 | - | 4 | 3534 | |||
AK134549.1 | - | 5 | 334 | |||
AK147670.1 | - | 5 | 383 | |||
AK160172.1 | - | 5 | 355 | |||
AK211128.1 | - | 1 | 23 | |||
AK213501.1 | - | 2 | 39 | |||
AK218333.1 | - | 2 | 39 | |||
BC047992.1 | - | 4 | 322 | 6504579 | ||
BC061045.1 | - | 5 | 395 | 6534037 |