Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054063.4 | NP_473404.3 | 2 | 422 | ||
GenBank mRNA | AF113598.1 | - | 2 | 272 | ||
BC099402.1 | - | 2 | 408 | 30787049 | ||
BC099403.1 | - | 2 | 293 | 30787292 | ||
BC141038.1 | - | 2 | 375 | 9055650 | ||
CK020635.1 | - | 2 | 422 |