Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psg28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054063.4 | NP_473404.3 | 3 | 842 | ||
GenBank mRNA | AF113598.1 | - | 3 | 692 | ||
BC099402.1 | - | 3 | 828 | 30787049 | ||
BC099403.1 | - | 3 | 713 | 30787292 | ||
BC141038.1 | - | 3 | 795 | 9055650 |