Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054070.3 | NP_473411.2 | 17 | 2240 | ||
GenBank mRNA | AF329695.1 | - | 17 | 2181 | ||
AK006708.1 | - | 11 | 1511 | |||
AK012394.1 | - | 3 | 417 | |||
AK032369.1 | - | 2 | 556 | |||
AK159647.1 | - | 17 | 2204 | |||
AK167964.1 | - | 17 | 2215 | |||
AK168244.1 | - | 16 | 2098 | |||
AK213495.1 | - | 2 | 113 | |||
BC043320.1 | - | 7 | 946 | |||
BC056978.1 | - | 17 | 2199 | 6821746 | ||
BC060064.1 | - | 16 | 2083 |