Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Afg3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054070.3 | NP_473411.2 | 2, 3 | 278 | ||
GenBank mRNA | AF329695.1 | - | 2, 3 | 219 | ||
AK014232.1 | - | 2, 3 | 201 | |||
AK159647.1 | - | 2, 3 | 242 | |||
AK167964.1 | - | 2, 3 | 253 | |||
AK168244.1 | - | 2, 3 | 252 | |||
BC056978.1 | - | 2, 3 | 237 | 6821746 | ||
BC060064.1 | - | 2, 3 | 237 |