Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rims1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012623.1 | NP_001012641.1 | 1983 | |||
NM_001012624.1 | NP_001012642.1 | 1983 | ||||
NM_001012625.1 | NP_001012643.1 | 1983 | ||||
NM_053270.1 | NP_444500.1 | 1983 | ||||
NM_183018.2 | NP_898839.2 | 1983 | ||||
GenBank mRNA | AK134597.1 | - | 894 | |||
AK220335.1 | - | 1916 | ||||
AY356534.1 | - | 1983 | ||||
AY356535.1 | - | 1983 | ||||
AY356536.1 | - | 1983 | ||||
AY356537.1 | - | 1983 | ||||
AY356538.1 | - | 1983 |