Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Iltifb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AJ249492.1 | - | 2, 3 | 302 |
Gene Symbol: Il22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016971.2 | NP_058667.1 | 2, 3 | 280 | ||
GenBank mRNA | BC116235.2 | - | 2, 3 | 276 | 40091712 |