Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mta3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001171052.1 | NP_001164523.1 | 13 | 1340 | ||
NM_001171053.1 | NP_001164524.1 | 13 | 1343 | |||
NM_001171054.1 | NP_001164525.1 | 13 | 1340 | |||
NM_054082.2 | NP_473423.1 | 13 | 1337 | |||
GenBank mRNA | AF288138.1 | - | 13 | 1291 | ||
AK003306.1 | - | 820 | ||||
AK129318.1 | - | 13 | 1272 | |||
AK145942.1 | - | 13 | 1285 | |||
AK146721.1 | - | 13 | 1293 | |||
AK146905.1 | - | 13 | 1293 | |||
AK147197.1 | - | 13 | 1283 | |||
AK154856.1 | - | 13 | 1280 | |||
AK158866.1 | - | 12 | 1203 | |||
BC022124.1 | - | 13 | 1304 | 5097283 | ||
BC099960.1 | - | 13 | 1260 | 30023887 | ||
CA751287.1 | - | 7 | 683 |