Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dph1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144491.2 | NP_652762.2 | 3 | 286 | ||
GenBank mRNA | AK082812.1 | - | 3 | 291 | ||
AK128996.1 | - | 277 | ||||
AK219998.1 | - | 2 | 206 | |||
AY078170.1 | - | 3 | 277 | |||
BB651982.1 | - | 3 | 291 | |||
BC044663.1 | - | 3 | 257 | |||
BC080761.1 | - | 3 | 262 |