Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tgs1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054089.4 | NP_473430.3 | 3 | 347 | ||
GenBank mRNA | AF389908.1 | - | 3 | 216 | ||
AK030953.1 | - | 3 | 339 | |||
AK044895.1 | - | 3 | 348 | |||
AK050044.1 | - | 3 | 277 | |||
AK170949.1 | - | 2 | 1820 | |||
BC058183.1 | - | 3 | 351 | |||
BC075728.1 | - | 3 | 328 | 6513964 |