Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam57a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027773.3 | NP_082049.1 | 3 | 460 | ||
GenBank mRNA | AK077374.1 | - | 3 | 420 | ||
AK160183.1 | - | 3 | 421 | |||
BC115519.1 | - | 3 | 459 | 40101189 | ||
BC115520.1 | - | 3 | 460 | 40101190 |