Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REG3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002580.2 | NP_002571.1 | 5 | 505 | ||
NM_138937.2 | NP_620354.1 | 5 | 482 | |||
NM_138938.2 | NP_620355.1 | 4 | 792 | |||
GenBank mRNA | AK291973.1 | - | 5 | 481 | ||
AY544128.1 | - | 4 | 529 | |||
AY544130.1 | - | 4 | 475 | |||
BC036776.1 | - | 4 | 486 | 5749980 | ||
BQ777112.1 | - | 5 | 505 | |||
D13510.1 | - | 4 | 481 | |||
M84337.1 | - | 4 | 486 | |||
S51768.1 | - | 4 | 486 | |||
X68641.1 | - | 4 | 457 |