Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc2a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012363.2 | NP_001012363.2 | 5 | 488 | ||
NM_001102414.1 | NP_001095884.1 | 5 | 687 | |||
NM_001102415.1 | NP_001095885.1 | 5 | 687 | |||
NM_145559.2 | NP_663534.1 | 6 | 682 | |||
GenBank mRNA | AF469480.1 | - | 5 | 691 | ||
AF490463.1 | - | 5 | 512 | |||
AK037093.1 | - | 5 | 540 | |||
AK156740.1 | - | 5 | 591 | |||
AK172234.1 | - | 5 | 591 | |||
AY776155.1 | - | 5 | 467 | |||
BC006076.1 | - | 6 | 682 | 3593581 | ||
BC138213.1 | - | 6 | 623 | 8861232 | ||
BC145198.1 | - | 6 | 623 | 9053749 |