Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002581.4 | NP_002572.2 | 17 | 4660 | ||
XM_006717129.2 | XP_006717192.1 | 15 | 2734 | |||
XM_017014784.1 | XP_016870273.1 | 16 | 4586 | |||
GenBank mRNA | AK300129.1 | - | 11 | 1591 | ||
BC078657.1 | - | 17 | 4290 | 30340336 | ||
X68280.1 | - | 17 | 4051 |