Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igf2bp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009951.4 | NP_034081.1 | 3 | 561 | ||
GenBank mRNA | AF061569.1 | - | 3 | 380 | ||
AK021257.1 | - | 2 | 122 | |||
AK044850.1 | - | 3 | 466 | |||
BC051679.1 | - | 3 | 544 | 30008106 |